Assay Details
Target Gene Details
Entrez Gene ID: | 17843 |
Gene Name: | major urinary protein 4 |
Gene Aliases: |
Mup-4, Mup1 |
Location: |
Chr.4:59956806-59960665 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mup4 | NM_008648.1 | NP_032674.1 | ||
| BC012227.1 | AAH12227.1 | |||
| M16358.1 | AAA39769.1 |
More Information
Set Membership: |
|

Genomic Map