Assay Details
Target Gene Details
Entrez Gene ID: | 100019 |
Gene Name: | midasin homolog (yeast) |
Gene Aliases: |
4833432B22Rik, A130070M06, AA958993, D4Abb1e, Gm135 |
Location: |
Chr.4:32657112-32775217 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdn1 | NM_001081392.1 | NP_001074861.1 | ||
| AK144722.1 |

Genomic Map