Assay Details
Target Gene Details
Entrez Gene ID: | 27354 |
Gene Name: | nibrin |
Gene Aliases: |
Nbs1 |
Location: |
Chr.4:15957967-15992589 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbn | NM_013752.3 | 7 | 910 | NP_038780.3 |
| AB016988.1 | 7 | 832 | BAA76298.1 | |
| AF076687.1 | 7 | 904 | AAD20943.1 | |
| AF092840.1 | 7 | 893 | AAC62113.1 | |
| AK031933.1 | 7 | 896 | BAC27610.1 | |
| AK132302.1 | 7 | 951 | ||
| AK134805.1 | 7 | 946 | ||
| AK134960.1 | 7 | 938 | BAE22356.1 | |
| AK159790.1 | 7 | 917 | ||
| AK160614.1 | 7 | 877 | ||
| AK183777.1 | 2 | 141 | ||
| BC044773.2 | 7 | 909 | AAH44773.1 | |
| BC055061.1 | 7 | 902 | AAH55061.1 |
More Information
Set Membership: |
|

Genomic Map