Assay Details
Target Gene Details
Entrez Gene ID: | 16157 |
Gene Name: | interleukin 11 receptor, alpha chain 1 |
Gene Aliases: |
AI314697, GP130, Il-11ra, Il11ra, Il11ra2, NR1 |
Location: |
Chr.4:41760443-41769473 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il11ra1 | NM_001163401.1 | NP_001156873.1 | ||
| NM_001172054.1 | NP_001165525.1 | |||
| NM_010549.3 | NP_034679.1 | |||
| AK143811.1 | ||||
| AK146742.1 | BAE27402.1 | |||
| AK155407.1 | BAE33248.1 | |||
| AK159559.1 | ||||
| BC004619.1 | AAH04619.1 | |||
| BC057664.1 | AAH57664.1 | |||
| BC069984.1 | AAH69984.1 | |||
| BY149476.1 | ||||
| BY777524.1 | ||||
| U14412.1 | AAA53248.1 | |||
| X74953.1 | CAA52908.1 |
More Information
Set Membership: |
|

Genomic Map