Assay Details
Target Gene Details
Entrez Gene ID: | 18201 |
Gene Name: | neutral sphingomyelinase (N-SMase) activation associated factor |
Gene Aliases: |
AA959567, C630007J05, Fan |
Location: |
Chr.4:6396207-6454668 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsmaf | NM_010945.2 | 28 | 2610 | NP_035075.2 |
| AF013632.1 | 28 | 2603 | AAB65394.1 | |
| AK172560.1 | BAE43067.1 | |||
| BC145961.1 | 28 | 2424 | ||
| BC145963.1 | 28 | 2424 |

Genomic Map