Assay Details
Target Gene Details
Entrez Gene ID: | 18201 |
Gene Name: | neutral sphingomyelinase (N-SMase) activation associated factor |
Gene Aliases: |
AA959567, C630007J05, Fan |
Location: |
Chr.4:6396207-6454668 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsmaf | NM_010945.2 | NP_035075.2 | ||
| AF013632.1 | AAB65394.1 | |||
| AK172560.1 | BAE43067.1 | |||
| BC145961.1 | ||||
| BC145963.1 | ||||
| CO043098.1 |

Genomic Map