Assay Details
Target Gene Details
Entrez Gene ID: | 101739 |
Gene Name: | PC4 and SFRS1 interacting protein 1 |
Gene Aliases: |
AA408851, AU015605, Dfs70, Ledgfa, Ledgfb, Psip2 |
Location: |
Chr.4:83455680-83486638 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psip1 | NM_001290527.1 | 1 | 77 | NP_001277456.1 |
| NM_133948.5 | 1 | 77 | NP_598709.1 | |
| AF339082.1 | 1 | 129 | AAO32949.1 | |
| AF339083.1 | 1 | 129 | AAO32950.1 | |
| AK011572.1 | BAB27707.3 | |||
| AK161426.1 | BAE36388.1 | |||
| BC027215.1 | 1 | 130 | ||
| BC043079.1 | AAH43079.1 | |||
| BC052177.1 | 1 | 80 | AAH52177.1 | |
| BY710824.1 | ||||
| CJ053610.1 | 1 | 78 |
More Information
Set Membership: |
|

Genomic Map