Assay Details
Target Gene Details
Entrez Gene ID: | 101739 |
Gene Name: | PC4 and SFRS1 interacting protein 1 |
Gene Aliases: |
AA408851, AU015605, Dfs70, Ledgfa, Ledgfb, Psip2 |
Location: |
Chr.4:83455680-83486638 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psip1 | NM_133948.5 | NP_598709.1 | ||
| AF339082.1 | AAO32949.1 | |||
| AJ308965.1 | CAC34944.1 | |||
| BC002260.1 | AAH02260.1 | |||
| BC043079.1 | AAH43079.1 |
Target Gene Details
Entrez Gene ID: | 77634 |
Gene Name: | small nuclear RNA activating complex, polypeptide 3 |
Gene Aliases: |
1810020H02Rik, 4930558A07Rik, 5031401C21Rik, AI414457, E030018J20Rik |
Location: |
Chr.4:83417701-83467658 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snapc3 | AK019863.2 | BAB31890.2 | ||
| AK031870.1 | BAC27585.1 | |||
| AK049933.1 | 10 | 2288 | BAC33990.1 |
More Information
Set Membership: |
|

Genomic Map