Assay Details
Target Gene Details
Entrez Gene ID: | 230514 |
Gene Name: | leptin receptor overlapping transcript |
Gene Aliases: |
LEPR, OB-RGRP, Obrgrp, VPS55 |
Location: |
Chr.4:101647783-101659358 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Leprot | NM_175036.4 | NP_778201.1 | ||
| AJ011565.1 | CAA09693.1 | |||
| AK009569.1 | BAB26366.1 | |||
| AK152008.1 | ||||
| AK159489.1 | BAE35124.1 | |||
| AK160453.1 | BAE35795.1 | |||
| AK165821.1 | BAE38395.1 | |||
| AK168597.1 | BAE40464.1 | |||
| BC004744.1 | AAH04744.1 | |||
| BC010289.1 | AAH10289.1 |
More Information
Set Membership: |
|

Genomic Map