Assay Details
Target Gene Details
Entrez Gene ID: | 68943 |
Gene Name: | PTEN induced putative kinase 1 |
Gene Aliases: |
1190006F07Rik, AU042772, AW557854, BRPK, mFLJ00387 |
Location: |
Chr.4:138313409-138326296 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pink1 | NM_026880.2 | NP_081156.2 | ||
| AB053476.1 | BAB55651.1 | |||
| AF316872.1 | AAK28061.1 | |||
| AK050322.1 | ||||
| AK155474.1 | BAE33284.1 | |||
| AK165480.1 | ||||
| AK180089.1 | ||||
| AK198589.1 | ||||
| AK220318.1 | BAD90392.1 | |||
| BC054349.1 | AAH54349.1 | |||
| BC067066.1 | AAH67066.1 | |||
| BC069968.1 | AAH69968.1 |
More Information
Set Membership: |
|

Genomic Map