Assay Details
Target Gene Details
Entrez Gene ID: | 67916 |
Gene Name: | phosphatidic acid phosphatase type 2B |
Gene Aliases: |
1110003O22Rik, 2610002D05Rik, AV025606, D4Bwg0538e, D4Bwg1535e, Lpp3, Ppab2b |
Location: |
Chr.4:105157347-105232767 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppap2b | NM_080555.2 | NP_542122.1 | ||
| AK011276.1 | ||||
| AK076183.1 | ||||
| AK159136.1 | BAE34848.1 | |||
| AK160056.1 | BAE35594.1 | |||
| BC005558.1 | AAH05558.1 |
More Information
Set Membership: |
|

Genomic Map