Assay Details
Target Gene Details
Entrez Gene ID: | 68350 |
Gene Name: | mitochondrial ubiquitin ligase activator of NFKB 1 |
Gene Aliases: |
0610009K11Rik, AV000801, Gide |
Location: |
Chr.4:138434661-138442265 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mul1 | NM_026689.3 | 1 | 149 | NP_080965.2 |
| AK002416.1 | 1 | 131 | BAB22084.1 | |
| AK044088.1 | 1 | 111 | BAC31768.1 | |
| AK076419.1 | 1 | 109 | BAC36332.1 | |
| AK083295.1 | 1 | 120 | BAC38848.1 | |
| AK153956.1 | 1 | 99 | BAE32279.1 | |
| AK170149.1 | 1 | 133 | BAE41597.1 | |
| BB856128.1 | 1 | 151 | ||
| BC019516.1 | 1 | 118 | AAH19516.1 | |
| BY062056.1 | 1 | 140 | ||
| EU935009.1 | 1 | 61 |

Genomic Map