Assay Details
Target Gene Details
Entrez Gene ID: | 71904 |
Gene Name: | progestin and adipoQ receptor family member VII |
Gene Aliases: |
2310021M12Rik, AV001994, BB007237, Mpra, PGLP, mPR |
Location: |
Chr.4:134496761-134510237 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paqr7 | NM_001285845.1 | NP_001272774.1 | ||
| NM_001285846.1 | NP_001272775.1 | |||
| NM_001285847.1 | NP_001272776.1 | |||
| NM_001285849.1 | NP_001272778.1 | |||
| NM_027995.3 | NP_082271.1 | |||
| AF313618.1 | AAO47231.1 | |||
| AK009450.1 | BAB26296.1 | |||
| AK078685.1 | ||||
| AK161542.1 | BAE36452.1 | |||
| AK170307.1 | BAE41704.1 | |||
| BY206669.1 | ||||
| BY210355.1 | ||||
| BY288540.1 |
More Information
Set Membership: |
|

Genomic Map