Assay Details
Target Gene Details
Entrez Gene ID: | 230577 |
Gene Name: | prolyl-tRNA synthetase (mitochondrial)(putative) |
Gene Aliases: |
BC027073 |
Location: |
Chr.4:106651021-106655282 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pars2 | NM_001083887.2 | NP_001077356.1 | ||
| NM_001285783.1 | NP_001272712.1 | |||
| NM_001285784.1 | NP_001272713.1 | |||
| NM_172272.2 | NP_758476.2 | |||
| AK154150.1 | BAE32409.1 | |||
| BB652093.1 | ||||
| BB842735.1 | ||||
| BC038817.1 | AAH38817.1 | |||
| BY768753.1 |
More Information
Set Membership: |
|

Genomic Map