Assay Details
Target Gene Details
Entrez Gene ID: | 17096 |
Gene Name: | Yamaguchi sarcoma viral (v-yes-1) oncogene homolog |
Gene Aliases: |
AA407514, Hck-2, p53Lyn, p56Lyn |
Location: |
Chr.4:3678121-3791612 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lyn | NM_001111096.1 | 1 | 168 | NP_001104566.1 |
| NM_010747.2 | 1 | 168 | NP_034877.2 | |
| AK028112.1 | 1 | 164 | BAC25753.1 | |
| AK151914.1 | 1 | 166 | ||
| AK152199.1 | 1 | 166 | BAE31028.1 | |
| AK153038.1 | 1 | 165 | BAE31669.1 | |
| AK156519.1 | 1 | 163 | ||
| AK170561.1 | 1 | 166 | BAE41882.1 | |
| AK172492.1 | 1 | 172 | ||
| BC003253.1 | 1 | 131 | ||
| BC031547.1 | 1 | 138 | AAH31547.1 | |
| BC157998.1 | ||||
| BY743506.1 | 1 | 173 | ||
| M57696.1 | AAA39471.1 | |||
| M57697.1 | AAA39472.1 | |||
| M64608.1 | 1 | 156 | AAA39470.1 |

Genomic Map