Assay Details
Target Gene Details
Entrez Gene ID: | 269513 |
Gene Name: | Na+/K+ transporting ATPase interacting 3 |
Gene Aliases: |
C530048M05, E130310K16Rik |
Location: |
Chr.4:20118336-20780310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkain3 | NM_001290410.1 | NP_001277339.1 | ||
| NM_172987.2 | NP_766575.2 | |||
| AK049776.1 | BAC33916.1 | |||
| AK053823.1 | BAC35541.1 | |||
| AK141456.1 | BAE24690.1 | |||
| BC075683.1 | AAH75683.1 | |||
| EF058052.1 | ||||
| EF058053.1 | ||||
| EF058054.1 |

Genomic Map