Assay Details
Target Gene Details
Entrez Gene ID: | 50793 |
Gene Name: | origin recognition complex, subunit 3 |
Gene Aliases: |
Orc3l |
Location: |
Chr.4:34566781-34614942 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orc3 | NM_001159563.1 | 5 | 433 | NP_001153035.1 |
| NM_015824.4 | 5 | 433 | NP_056639.3 | |
| AB190257.2 | 5 | 400 | BAD91665.2 | |
| AJ132830.1 | 5 | 415 | CAB76399.1 | |
| AK045388.1 | 5 | 408 | BAC32339.1 | |
| AK086579.1 | 5 | 436 | BAC39695.1 | |
| AK145324.1 | 5 | 412 | BAE26368.1 | |
| BB663298.1 | 5 | 433 | ||
| BC061252.1 | 5 | 404 | AAH61252.1 |
More Information
Set Membership: |
|

Genomic Map