Assay Details
Target Gene Details
Entrez Gene ID: | 230082 |
Gene Name: | nucleolar protein family 6 (RNA-associated) |
Gene Aliases: |
AA410091, Nrap |
Location: |
Chr.4:41114427-41124339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol6 | NM_139236.3 | NP_631982.2 | ||
| AF361077.1 | AAL74401.1 | |||
| AF361078.1 | AAL74402.1 | |||
| AK089138.1 | ||||
| AK155818.1 | BAE33447.1 | |||
| AK166559.1 | ||||
| AK166650.1 | BAE38917.1 | |||
| AK171163.1 | BAE42285.1 | |||
| AK171421.1 | BAE42443.1 | |||
| AK189927.1 | ||||
| AK203163.1 | ||||
| AK205172.1 | ||||
| BC059820.1 |
More Information
Set Membership: |
|

Genomic Map