Assay Details
Target Gene Details
Entrez Gene ID: | 381511 |
Gene Name: | pyruvate dehyrogenase phosphatase catalytic subunit 1 |
Gene Aliases: |
Gm1024, Ppm2c |
Location: |
Chr.4:11958183-11966450 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdp1 | NM_001033453.3 | NP_001028625.1 | ||
| NM_001098230.1 | NP_001091700.1 | |||
| AK133542.1 | ||||
| AK137550.1 | BAE23403.1 | |||
| BB611000.1 | ||||
| BY101287.1 | ||||
| CJ147172.1 |
More Information
Set Membership: |
|

Genomic Map