Assay Details
Target Gene Details
Entrez Gene ID: | 22004 |
Gene Name: | tropomyosin 2, beta |
Gene Aliases: |
Tpm-2, Trop-2 |
Location: |
Chr.4:43513726-43523583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpm2 | NM_001277875.1 | NP_001264804.1 | ||
| NM_001277876.1 | NP_001264805.1 | |||
| NM_009416.4 | NP_033442.2 | |||
| AA208641.1 | ||||
| AK003186.1 | BAB22629.1 | |||
| AK132162.1 | ||||
| BC014809.1 | AAH14809.1 | |||
| BC024358.1 | AAH24358.1 | |||
| BY439632.1 | ||||
| DV049735.1 | ||||
| M81086.1 | AAA40484.1 | |||
| M87635.1 | AAA37288.1 | |||
| X12650.1 | CAA31181.1 | |||
| X58381.1 | CAA41271.1 |
More Information
Set Membership: |
|

Genomic Map