Assay Details
Target Gene Details
Entrez Gene ID: | 269523 |
Gene Name: | valosin containing protein |
Gene Aliases: |
3110001E05, CDC48, p97, p97/VCP |
Location: |
Chr.4:42979964-43000507 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |

Genomic Map