Assay Details
Target Gene Details
Entrez Gene ID: | 269523 |
Gene Name: | valosin containing protein |
Gene Aliases: |
3110001E05, CDC48, p97, p97/VCP |
Location: |
Chr.4:42979964-43000507 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vcp | NM_009503.4 | NP_033529.3 | ||
| AK028264.1 | BAC25849.1 | |||
| AK030751.1 | BAC27119.1 | |||
| AK083821.1 | BAC39028.1 | |||
| AK146109.1 | ||||
| AK149931.1 | BAE29175.1 | |||
| AK151109.1 | BAE30119.1 | |||
| AK151418.1 | BAE30383.1 | |||
| AK153012.1 | ||||
| AK153249.1 | BAE31840.1 | |||
| AK159177.1 | BAE34876.1 | |||
| AK159475.1 | ||||
| AK159509.1 | BAE35141.1 | |||
| AK160889.1 | ||||
| AK167794.1 | BAE39824.1 | |||
| AK169140.1 | BAE40919.1 | |||
| AK183217.1 | ||||
| AK185530.1 | ||||
| AK187846.1 | ||||
| AK189857.1 | ||||
| AK193877.1 | ||||
| AK193943.1 | ||||
| AK195731.1 | ||||
| AK198927.1 | ||||
| AK203529.1 | ||||
| AK209970.1 | ||||
| AK210550.1 | ||||
| AK211047.1 | ||||
| AK211479.1 | ||||
| AK213510.1 | ||||
| AK213824.1 | ||||
| AK214199.1 | ||||
| AK214818.1 | ||||
| AK216568.1 | ||||
| AK217520.1 | ||||
| AK218438.1 | ||||
| AK218772.1 | ||||
| BC043053.1 | AAH43053.1 | |||
| BC049114.1 | AAH49114.1 | |||
| Z14044.1 | CAA78412.1 |

Genomic Map