Assay Details
Target Gene Details
Entrez Gene ID: | 242466 |
Gene Name: | zinc finger protein 462 |
Gene Aliases: |
6030417H05, 9430078C22Rik, Gt4-2, Zfpip |
Location: |
Chr.4:54944583-55083563 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp462 | NM_172867.3 | NP_766455.2 | ||
| AK173266.1 | BAD32544.1 | |||
| BC099692.1 | AAH99692.1 | |||
| DQ355518.1 | ||||
| X66902.1 | CAA47353.1 |
More Information
Set Membership: |
|

Genomic Map