Assay Details
Target Gene Details
Entrez Gene ID: | 27096 |
Gene Name: | trafficking protein particle complex 3 |
Gene Aliases: |
1110058K12Rik, AI835942, AU019197, Bet3 |
Location: |
Chr.4:126262405-126275883 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc3 | NM_013718.2 | NP_038746.1 | ||
| AF041433.1 | AAB96937.1 | |||
| AK151747.1 | BAE30657.1 | |||
| AK168518.1 | ||||
| BC003736.1 | AAH03736.1 |

Genomic Map