Assay Details
Target Gene Details
Entrez Gene ID: | 72701 |
Gene Name: | zinc finger protein 618 |
Gene Aliases: |
2810031P15Rik, 2810040O04Rik, AI415580, AI505139, D430033D05Rik, Nedd10, Znf618, bM257P7.1 |
Location: |
Chr.4:62965555-63139709 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp618 | NM_028326.1 | NP_082602.1 | ||
| AK012848.1 | BAB28510.1 |
More Information
Set Membership: |
|

Genomic Map