Assay Details
Target Gene Details
Entrez Gene ID: | 74255 |
Gene Name: | smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans) |
Gene Aliases: |
2600001O03Rik, 2610203K23Rik, AB044414, AI845086, AW556129, Bwd |
Location: |
Chr.4:40735649-40757885 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smu1 | NM_021535.4 | NP_067510.3 | ||
| AB044414.1 | BAA96656.1 | |||
| AK003493.1 | BAB22820.1 | |||
| AK011140.1 | 8 | 1052 | BAC25322.1 | |
| AK019174.1 | ||||
| AK145982.1 | BAE26804.1 | |||
| AK159635.1 | BAE35249.1 | |||
| AK168813.1 | BAE40640.1 | |||
| BC057446.1 | AAH57446.1 |

Genomic Map