Assay Details
Target Gene Details
Entrez Gene ID: | 77634 |
Gene Name: | small nuclear RNA activating complex, polypeptide 3 |
Gene Aliases: |
1810020H02Rik, 4930558A07Rik, 5031401C21Rik, AI414457, E030018J20Rik |
Location: |
Chr.4:83417701-83467658 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snapc3 | NM_029949.3 | NP_084225.1 | ||
| AK007563.1 | ||||
| AK010932.1 | BAB27276.1 | |||
| AK016168.1 | BAB30136.1 | |||
| AK019863.2 | BAB31890.2 | |||
| AK031870.1 | BAC27585.1 | |||
| AK031924.1 | ||||
| AK049933.1 | BAC33990.1 | |||
| BC048713.1 | AAH48713.1 |
More Information
Set Membership: |
|

Genomic Map