Assay Details
Target Gene Details
Entrez Gene ID: | 22249 |
Gene Name: | unc-13 homolog B (C. elegans) |
Gene Aliases: |
Munc13-1, Munc13-2, Unc13a, Unc13h1, Unc13h2 |
Location: |
Chr.4:43058862-43264887 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc13b | NM_001081413.1 | 15 | 1801 | NP_001074882.1 |
| NM_021468.2 | 15 | 1712 | NP_067443.2 | |
| AF115848.1 | 14 | 1783 | AAD13619.1 | |
| AK122422.1 | 9 | 1991 | BAC65704.3 | |
| BC157967.1 | 15 | 1712 | ||
| BC158025.1 | 15 | 1712 |

Genomic Map