Assay Details
Target Gene Details
Entrez Gene ID: | 116940 |
Gene Name: | trimethylguanosine synthase homolog (S. cerevisiae) |
Gene Aliases: |
D4Ertd800e, Ncoa6ip, Pimt |
Location: |
Chr.4:3574879-3616624 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgs1 | NM_054089.4 | NP_473430.3 | ||
| AF389908.1 | AAK84355.1 | |||
| AK030953.1 | ||||
| AK044895.1 | ||||
| BC026368.1 | AAH26368.1 | |||
| BC075728.1 | AAH75728.1 |
More Information
Set Membership: |
|

Genomic Map