Assay Details
Target Gene Details
Entrez Gene ID: | 100041581 |
Gene Name: | zinc finger with KRAB and SCAN domains 16 |
Gene Aliases: |
XM_143800, Zfp483 |
Location: |
Chr.4:58943568-58962624 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zkscan16 | NM_001099323.2 | NP_001092793.1 |
