Assay Details
Target Gene Details
Entrez Gene ID: | 72519 |
Gene Name: | transmembrane protein 55A |
Gene Aliases: |
2610319K07Rik, AI315591, AV001360 |
Location: |
Chr.4:14864219-14915260 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem55a | NM_028264.4 | NP_082540.1 | ||
| AK012054.1 | BAB27995.1 | |||
| AK033845.1 | BAC28493.1 | |||
| AK049213.1 | BAC33613.1 | |||
| AK185686.1 | ||||
| AK206695.1 | ||||
| AX775959.1 | CAE11679.1 | |||
| BC021435.1 | AAH21435.1 |

Genomic Map