Assay Details
Target Gene Details
Entrez Gene ID: | 60599 |
Gene Name: | transformation related protein 53 inducible nuclear protein 1 |
Gene Aliases: |
2700057G22Rik, SIP, SIP18, SIP27, Stinp, Teap |
Location: |
Chr.4:11156441-11174377 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53inp1 | NM_001199105.1 | NP_001186034.1 | ||
| NM_021897.3 | NP_068697.1 | |||
| AJ131776.1 | CAB66138.1 | |||
| AK054214.1 | ||||
| AK142321.1 | BAE25030.1 | |||
| AY034611.1 | AAK60419.1 | |||
| AY034612.1 | AAK60420.1 |

Genomic Map