Assay Details
Target Gene Details
Entrez Gene ID: | 22590 |
Gene Name: | xeroderma pigmentosum, complementation group A |
Gene Aliases: |
AI573865, Xpac |
Location: |
Chr.4:46174746-46196349 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xpa | NM_011728.2 | NP_035858.2 | ||
| AF354052.1 | AAL83963.1 | |||
| AI172779.1 | ||||
| AI838722.1 | ||||
| AK008962.1 | BAB25992.1 | |||
| BP771655.1 | ||||
| X74351.1 | CAA52393.1 |
More Information
Set Membership: |
|

Genomic Map