Assay Details
Target Gene Details
Entrez Gene ID: | 74438 |
Gene Name: | clavesin 1 |
Gene Aliases: |
4933402J24Rik, Clvl1, Rlbp1l1 |
Location: |
Chr.4:9144756-9451691 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clvs1 | NM_028940.2 | NP_083216.1 | ||
| AK016622.1 | BAB30342.1 | |||
| AK032549.1 | ||||
| AK084350.1 | BAC39166.1 | |||
| AK136083.1 | BAE22811.1 | |||
| BC062923.1 | AAH62923.1 | |||
| BC100556.1 | AAI00557.1 |
More Information
Set Membership: |
|

Genomic Map