Assay Details
Target Gene Details
Entrez Gene ID: | 74438 |
Gene Name: | clavesin 1 |
Gene Aliases: |
4933402J24Rik, Clvl1, Rlbp1l1 |
Location: |
Chr.4:9144756-9451691 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clvs1 | NM_028940.2 | 5 | 1191 | NP_083216.1 |
| AK016622.1 | 5 | 1115 | BAB30342.1 | |
| AK032549.1 | 5 | 1181 | ||
| AK084350.1 | 5 | 1187 | BAC39166.1 | |
| AK136083.1 | 4 | 726 | BAE22811.1 | |
| BC062923.1 | 5 | 1146 | AAH62923.1 | |
| BC100556.1 | 3 | 411 | AAI00557.1 |
More Information
Set Membership: |
|

Genomic Map