Assay Details
Target Gene Details
Entrez Gene ID: | 66625 |
Gene Name: | PNN interacting serine/arginine-rich |
Gene Aliases: |
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18 |
Location: |
Chr.4:21847583-21876475 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnisr | NM_025669.1 | 10 | 1767 | NP_079945.1 |
| AK012036.1 | 10 | 1309 | ||
| AK017507.1 | 10 | 1294 | BAB30779.3 | |
| AK020035.1 | 10 | 1300 | ||
| AK031964.1 | 10 | 1303 | ||
| AK077040.1 | 8 | 3016 | ||
| AK160288.1 | 10 | 1295 | BAE35727.1 | |
| BC141031.1 | 10 | 1631 |

Genomic Map