Assay Details
Target Gene Details
Entrez Gene ID: | 66625 |
Gene Name: | PNN interacting serine/arginine-rich |
Gene Aliases: |
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18 |
Location: |
Chr.4:21847583-21876475 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnisr | NM_025669.1 | 4 | 954 | NP_079945.1 |
| AK012036.1 | 4 | 497 | ||
| AK017507.1 | 4 | 481 | BAB30779.3 | |
| AK020035.1 | 4 | 482 | ||
| AK031964.1 | 4 | 490 | ||
| AK077040.1 | 3 | 678 | ||
| AK081407.1 | 3 | 2195 | ||
| AK083021.1 | 4 | 480 | ||
| AK160288.1 | 4 | 480 | BAE35727.1 | |
| AK162695.1 | 4 | 507 | ||
| BC139811.1 | 1 | 105 | ||
| BC141031.1 | 4 | 818 |

Genomic Map