Assay Details
Target Gene Details
Entrez Gene ID: | 230734 |
Gene Name: | yrdC domain containing (E.coli) |
Gene Aliases: |
AV303379, BC023823, IRIP, ITIP |
Location: |
Chr.4:124850759-124855242 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yrdc | NM_153566.2 | 3 | 586 | NP_705794.2 |
| AK153627.1 | 3 | 586 | BAE32125.1 | |
| AK219737.1 | 1 | 51 | ||
| AY283537.1 | 3 | 562 | AAP37032.1 | |
| BC023823.1 | 3 | 572 | AAH23823.1 |

Genomic Map