Assay Details
Target Gene Details
Entrez Gene ID: | 230734 |
Gene Name: | yrdC domain containing (E.coli) |
Gene Aliases: |
AV303379, BC023823, IRIP, ITIP |
Location: |
Chr.4:124850759-124855242 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yrdc | NM_153566.2 | NP_705794.2 | ||
| AK153627.1 | BAE32125.1 | |||
| AK219737.1 | ||||
| AY283537.1 | AAP37032.1 | |||
| BC023823.1 | AAH23823.1 |

Genomic Map