Assay Details
Target Gene Details
Entrez Gene ID: | 109093 |
Gene Name: | arginyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
1500002I10Rik, PRO1992, Rarsl |
Location: |
Chr.4:34614941-34660167 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rars2 | NM_181406.3 | NP_852071.2 | ||
| AK075721.1 | ||||
| AK156182.1 | BAE33614.1 | |||
| AK166986.1 | ||||
| BC024878.1 | AAH24878.1 |
More Information
Set Membership: |
|

Genomic Map