Assay Details
Target Gene Details
Entrez Gene ID: | 52187 |
Gene Name: | Ras-related GTP binding D |
Gene Aliases: |
5730543C08Rik, AI467523, C030003H22Rik, D4Ertd174e |
Location: |
Chr.4:32982998-33022180 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rragd | NM_027491.2 | NP_081767.2 | ||
| AK017818.1 | BAB30954.2 | |||
| BC038137.1 | AAH38137.1 |
More Information
Set Membership: |
|

Genomic Map