Assay Details
Target Gene Details
Entrez Gene ID: | 72098 |
Gene Name: | transmembrane protein 68 |
Gene Aliases: |
2010300G19Rik, AA408325, AW123402 |
Location: |
Chr.4:3549041-3574848 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem68 | NM_028097.3 | NP_082373.1 | ||
| AK008493.1 | BAB25698.1 | |||
| AK048210.1 | BAC33273.1 | |||
| AK084881.1 | BAC39301.1 | |||
| AK088600.1 | 1 | 37 | BAC40445.1 | |
| AK147765.1 | BAE28123.1 | |||
| AK162563.1 | BAE36969.1 | |||
| AK165916.1 | 1 | 56 | BAE38459.1 | |
| AK166237.1 | 1 | 80 | BAE38652.1 | |
| BC016240.1 | AAH16240.1 |

Genomic Map