Assay Details
Target Gene Details
Entrez Gene ID: | 24060 |
Gene Name: | solute carrier family 35 (CMP-sialic acid transporter), member 1 |
Gene Aliases: |
AA408150, AI314851 |
Location: |
Chr.4:34663257-34687812 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35a1 | NM_011895.3 | 7 | 1003 | NP_036025.2 |
| AK144170.1 | 7 | 996 | ||
| BC012252.1 | 7 | 918 | AAH12252.1 | |
| Z71268.1 | 7 | 824 | CAA95855.1 |
More Information
Set Membership: |
|

Genomic Map