Assay Details
Target Gene Details
Entrez Gene ID: | 21885 |
Gene Name: | transducin-like enhancer of split 1, homolog of Drosophila E(spl) |
Gene Aliases: |
C230057C06Rik, Estm14, Grg-1, Grg1, Tle4l |
Location: |
Chr.4:72109945-72200893 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tle1 | NM_001285529.1 | NP_001272458.1 | ||
| NM_001285530.1 | NP_001272459.1 | |||
| NM_011599.5 | NP_035729.3 | |||
| AK048515.1 | ||||
| AK052961.1 | BAC35221.1 | |||
| AK082499.1 | BAC38509.1 | |||
| AY155195.1 | AAN77514.1 | |||
| AY155197.1 | AAN77516.1 | |||
| AY155198.1 | AAN77517.1 | |||
| AY155199.1 | AAN77518.1 | |||
| AY155200.1 | AAN77519.1 | |||
| BC057573.1 | AAH57573.1 | |||
| U61362.1 | AAB49934.1 |
More Information
Set Membership: |
|

Genomic Map