Assay Details
Target Gene Details
Entrez Gene ID: | 20404 |
Gene Name: | SH3-domain GRB2-like 2 |
Gene Aliases: |
9530001L19Rik, AI120490, AW555077, B930049H17Rik, EEN-B1, SH3PA, Sh3d2a |
Location: |
Chr.4:85205456-85389380 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3gl2 | NM_019535.2 | NP_062408.2 | ||
| AF326561.1 | AAL37407.1 | |||
| AF326562.1 | AAL37408.1 | |||
| AK044370.1 | BAC31888.1 | |||
| AK134970.1 | BAE22364.1 | |||
| AK139337.1 | BAE23963.1 | |||
| BC018385.1 | AAH18385.1 | |||
| U58886.1 | AAC71774.1 |

Genomic Map