Assay Details
Target Gene Details
Entrez Gene ID: | 21923 |
Gene Name: | tenascin C |
Gene Aliases: |
AI528729, C130033P17Rik, Hxb, TN, TN-C, Ten, cytotactin, tenascin-C |
Location: |
Chr.4:63959785-64047015 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnc | NM_011607.3 | NP_035737.2 | ||
| AK085667.1 | ||||
| BC117979.1 | ||||
| BC117980.1 | ||||
| CN692948.1 | ||||
| D90343.1 | BAA14355.1 | |||
| X56304.1 | CAA39751.1 |
More Information
Set Membership: |
|

Genomic Map