Assay Details
Target Gene Details
Entrez Gene ID: | 381629 |
Gene Name: | all-trans retinoic acid induced differentiation factor |
Gene Aliases: |
0610007C21Rik, AI316792, Apr3, HSPC013, p18 |
Location: |
Chr.5:31048640-31054633 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atraid | NM_027855.4 | NP_082131.2 | ||
| NM_212470.3 | NP_997635.1 | |||
| AB009018.1 | ||||
| AK002276.1 | BAB21981.1 | |||
| AK151307.1 | BAE30289.1 | |||
| AK151366.1 | BAE30340.1 | |||
| AK158882.1 | ||||
| AK169267.1 | BAE41028.1 | |||
| AW987473.1 | ||||
| BC049637.1 | AAH49637.1 | |||
| BC057097.1 | AAH57097.2 |

Genomic Map