Assay Details
Target Gene Details
Entrez Gene ID: | 63954 |
Gene Name: | retinol binding protein 7, cellular |
Gene Aliases: |
1110002J23Rik, CRBP-III |
Location: |
Chr.4:149449687-149466462 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbp7 | NM_022020.2 | 4 | 492 | NP_071303.1 |
| AF260923.1 | 4 | 493 | AAG17284.1 | |
| AK003307.1 | 3 | 300 | BAB22705.1 | |
| AY141971.1 | 4 | 502 | AAN34937.1 | |
| BC028432.1 | 4 | 489 | AAH28432.1 |

Genomic Map