Assay Details
Target Gene Details
Entrez Gene ID: | 100213 |
Gene Name: | RUN and SH3 domain containing 2 |
Gene Aliases: |
AI840675, mKIAA0375 |
Location: |
Chr.4:43381982-43427092 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rusc2 | NM_001037709.2 | NP_001032798.1 | ||
| NM_199057.3 | 1 | 121 | NP_951012.2 | |
| AK053542.1 | BAC35421.1 | |||
| AK122263.1 | 1 | 43 | BAC65545.1 | |
| AK132170.1 | 1 | 50 | BAE21009.1 | |
| AK143020.1 | 1 | 1577 | ||
| AK166260.1 | ||||
| BC050780.1 | AAH50780.1 | |||
| BY237595.1 | 1 | 122 | ||
| BY283112.1 | ||||
| BY283239.1 |

Genomic Map