Assay Details
Target Gene Details
Entrez Gene ID: | 100213 |
Gene Name: | RUN and SH3 domain containing 2 |
Gene Aliases: |
AI840675, mKIAA0375 |
Location: |
Chr.4:43381982-43427092 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rusc2 | NM_001037709.2 | 5 | 3207 | NP_001032798.1 |
| NM_199057.3 | 5 | 3168 | NP_951012.2 | |
| AK053542.1 | 2 | 1469 | BAC35421.1 | |
| AK122263.1 | 5 | 3087 | BAC65545.1 | |
| AK132170.1 | 5 | 3097 | BAE21009.1 | |
| AK166260.1 | 5 | 3108 | ||
| BC024790.1 | 3 | 398 | AAH24790.1 | |
| BC141186.1 | 4 | 2938 |

Genomic Map